Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTIP Chicken anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™

Catalog No. ABE69MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABE69MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABE69MI Supplier MilliporeSigma Supplier No. ABE69
Add to Cart
Add to Cart

Chicken Polyclonal Antibody

Anti-PTimmunoprecipitation Antibody is a Chicken Polyclonal Antibody for detection of PTimmunoprecipitation also known as PAX transactivation activation domain-interacting protein, protein encoded by CAG trinucleotide repeats and has been validated in western blot, immunohistochemistry and immunofluoresence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTIP
Applications Immunofluorescence, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified chicken polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Symbols PAXIP1L; PTIP; PAXIP1; TNRC2; CAGF28; PACIP1; CAGF29
Host Species Chicken
Immunogen Histidine-tagged recombinant protein corresponding to human PTIP.
Purification Method Affinity Purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NP_031375
Target Species Human, Mouse, Rat
Content And Storage 2-8°C for 12 months
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.