Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PTP mu/PTPRM Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NB10079867
Description
PTP mu/PTPRM Polyclonal specifically detects PTP mu/PTPRM in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.Specifications
PTP mu/PTPRM | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
EC 3.1.3.48, hR-PTPu, MGC166994, protein tyrosine phosphatase mu, protein tyrosine phosphatase, receptor type, M, protein tyrosine phosphatase, receptor type, mu polypeptide, Protein-tyrosine phosphatase mu, PTPRL1R-PTP-MU, receptor-type tyrosine-protein phosphatase mu, RPTPM, R-PTP-mu, RPTPU | |
Goat | |
Affinity Purified | |
RUO | |
Primary | |
This antibody is expected to recognize both reported isoforms (NP_001098714.1 and NP_002836.3). | |
Store at -20C. Avoid freeze-thaw cycles. |
Immunohistochemistry (Paraffin), ELISA | |
0.5 mg/mL | |
Immunohistochemistry-Paraffin, Peptide ELISA Detection limit 1:16000 | |
NP_001098714.1 | |
PTPRM | |
Peptide with sequence C-KLIRQVDKWQEEYN corresponding to internal region according to NP_001098714.1, NP_002836.3. | |
0.1 mg | |
Protein Phosphatase | |
5797 | |
Human, Mouse, Canine | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction