Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-PTP, T Cell Clone: CF4-1D,
Mouse Monoclonal Antibody
Supplier: MilliporeSigma™ PH03L100UG
Description
Specifically detects PTP, T Cell Clone: CF4-1D in Human samples, and it is validated for Immunocytochemistry, Immunoprecipitation
Protein tyrosine phosphatases (PTPases) make up an expanding family of transmembrane and intracellular proteins, which function to remove phosphate groups from protein phosphoryl- tyrosine residues. The prototype phosphatase, PTPase 1B, was initially identified as an active intracellular phosphatase from human placenta made up of 321 amino acids and subsequently shown to derive from a larger precursor protein containing 435 amino acids. Using probes to the conserved PTPase domain a cDNA clone was isolated from a human peripheral T cell library encoding a protein with 65% identity to the placental PTPase 1B. The T cell protein differs in amino acid composition but is highly similar with respect to hydrophobicity with the 1B protein and is now known to be present in other cell types. While the function of the T cell PTPase protein is still unknown, recent experiments demonstrated that a truncated form overexpressed in BHK cells resulted in a greatly reduced growth rate of the cells. PTPases are involved in regulating processes where protein kinases and protein tyrosine phosphorylation are important. One intriguing hypothesis is that PTPases have the potential to function as anti-oncogenes.
Specifications
PTP, T Cell | |
Monoclonal | |
Unconjugated | |
Mouse | |
100 μg | |
Primary | |
Human | |
Purified |
Immunocytochemistry, Immunoprecipitation | |
CF4-1D | |
Lyophilized from 20mM sodium bicarbonate, 100μg BSA. | |
recombinant, human T cell PTPase | |
RUO | |
Recognizes the T-cell PTPase protein. | |
2°C to 8°C | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction