Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTP1B Mouse, Unlabeled, Clone: 15, BD

Catalog No. BDB610139 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610139 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB610139 Supplier BD Biosciences Supplier No. 610139
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PTP1B is a member of the class of enzymes termed protein tyrosine phosphatases(PTPs). This 50 kDa protein contains a conserved phosphatases domain at residues 30-278 and is localized to the cytoplasmic face of the endoplasmic reticulum by its C-terminal 35 residues. Its interactions with other proteins are mediated by proline-rich regions and SH2 compatible sequences. PTP1B is thought to act as a negative regulator in insulin signaling. Insulin stimulation induces its association with the insulin receptor and tyrosine phosphorylation. Its overexpression inhibits proximal and distal insulin signaling events, possibly due to receptor dephosphorylation. Microinjection of PTP1B into Xenopus oocytes blocks insulin-induced maturation. Additional studies demonstrate that PTP1B undergoes cell cycle-dependent and/or stress-induced serine phosphorylation. This suggests further regulatory roles that are not yet defined.

TRUSTED_SUSTAINABILITY

Specifications

Antigen PTP1B
Applications Western Blot
Classification Monoclonal
Clone 15
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Chicken, Frog, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG2a
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.