Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTPN13/PTPL1 Mouse anti-Human, Clone: 359313, R&D Systems™

Catalog No. MAB3577
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3577 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB3577 Supplier R&D Systems Supplier No. MAB3577
This item is not returnable. View return policy

Mouse Monoclonal Antibody

PTPN13/PTPL1 Monoclonal antibody MAB3577 Specifically detects PTPN13/PTPL1 in Human samples. It is validated for Western Blotting

Specifications

Antigen PTPN13/PTPL1
Applications Western Blot
Classification Monoclonal
Clone 359313
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q12923
Gene Alias EAP-1, FAP-1, hPTPE1, PNP1, protein tyrosine phosphatase, non-receptor type 13 (APO-1/CD95 (Fas)-associatedphosphatase), protein-tyrosine phosphatase 1, Fas-associated, PTP1E, PTP1EDKFZp686J1497, PTP-BAS, PTP-BASEC 3.1.3.48, PTP-BL, PTP-E1, PTPL1, PTPL1nonreceptor type 13, PTPLE, tyrosine-protein phosphatase non-receptor type 13
Gene Symbols PTPN13
Host Species Mouse
Immunogen E. coli-derived recombinant human PTPN13/PTPL1, His2-Arg500, Accession # Q12923
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5783
Test Specificity Detects endogenous human PTPN13/PTPL1 in Western blots.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.