Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
protein tyrosine phosphatase, nonreceptor type 5 (striatum-enriched), Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human PTPN5 protein.
Supplier: Abnova Corporation H00084867B01
Description
Sequence: MNYEGARSERENHAADDSEGGALDMCCSERLPGLPQPIVMEALDEAEGLQDSQREMPPPPPPSPPSDPAQKPPPRGAGSHSLTVRSSLCLFAASQFLLACGVLWFSGYGHIWSQNATNLVSSLLTLLKQLEPTAWLDSGTWGVPSLLLVFLSVGLVLVTTLVWHLLRTPPEPPTPLPPEDRRQSVSRQPSFTYSEWMEEKIEDDFLDLDPVPETPVFDCVMDIKPEADPTSLTVKSMGLQERRGSNVSLTLDMCTPGCNEEGFGYLMSPREESAREYLLSASRVLQAEELHEKALDPFLLQAEFFEIPMNFVDPKEYDIPGLVRKNRYKTILPNPHSRVCLTSPDPDDPLSSYINANYIRGYGGEEKVYIATQGPIVSTVADFWRMVWQEHTPIIVMITNIEEMNEKCTEYWPEEQVAYDGVEITVQKVIHTEDYRLRLISLKSGTEERGLKHYWFTSWPDQKTPDRAPPLLHLVREVEEAAQQEGPHCAPIIVHCSAGIGRTGCFIATSICCQQLRQEGVVDILKTTCQLRQDRGGMIQTCEQYQFVHHVMSLYEKQLSHQSPESpecifications
protein tyrosine phosphatase, non-receptor type 5 (striatum-enriched) | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human PTPN5 protein. | |
PTPN5 | |
FLJ14427/PTPSTEP/STEP | |
Mouse | |
50 μL | |
Yes | |
84867 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Western Blot | |
Unconjugated | |
No additive | |
NM_006906.1 | |
PTPN5 | |
PTPN5 (NP_008837.1, 1 a.a. ∼ 565 a.a) full-length human protein. | |
RUO | |
Primary | |
Human | |
Serum |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction