Learn More
signal-regulatory protein alpha, Mouse, Clone: 4C7, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant SIRPA.
Supplier: Abnova Corporation H00140885M10
Description
The protein encoded by this gene is a member of the signal-regulatory-protein (SIRP) family, and also belongs to the immunoglobulin superfamily. SIRP family members are receptor-type transmembrane glycoproteins known to be involved in the negative regulation of receptor tyrosine kinase-coupled signaling processes. This protein can be phosphorylated by tyrosine kinases. The phospho-tyrosine residues of this PTP have been shown to recruit SH2 domain containing tyrosine phosphatases (PTP), and serve as substrates of PTPs. This protein was found to participate in signal transduction mediated by various growth factor receptors. CD47 has been demonstrated to be a ligand for this receptor protein. This gene and its product share very high similarity with several other members of the SIRP family. These related genes are located in close proximity to each other on chromosome 20p13. Multiple alternatively spliced transcript variants have been determined for this gene. [provided by RefSeq
Sequence: VAGEEELQVIQPDKSVSVAAGESAILHCTVTSLIPVGPIQWFRGAGPARELIYNQKEGHFPRVTTVSESTKRENMDFSISISNITPADAGTYYCVKFRKGSPDTEFKSGAGTELSVRAKPSAPVVSGPAARATPQHTVSFTCESHGFSPRDITLKWFKNGNELSDFQTNVDPVGESVSYSIHSTAKVVLTREDVHSQVICEVAHVTLQGDPLRGTANLSETIRVPPTLEVTQQPVRAENQVNVTCQVRKFYPQRLQLTWLENGNVSRTETASTVTENKDGTYNWMSWLLVNVSAHRDDVKLTCQVEHDGQPAVSKSHDLKVSAHPKEQGSNTAAENTGSNERNIYIVVGVVCTLLVALLMAALYLVRIRQKKAQGSTSSTRLHEPEKNAREITQVQSLDTNDITYADLNLPKGKKPAPQAAEPNNHTEYASIQTSPQPASEDTLTYADLDMVHLNRTPKQPAPKPEPSFSEYASVQVPRKSpecifications
signal-regulatory protein alpha | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
BC026692 | |
SIRPA | |
SIRPA (AAH26692, 28 a.a. ∼ 507 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Yes | |
140885 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG2a κ |
ELISA, Immunofluorescence, Western Blot | |
4C7 | |
Mouse monoclonal antibody raised against a full length recombinant SIRPA. | |
SIRPA | |
BIT/CD172A/MFR/MYD-1/P84/PTPNS1/SHPS-1/SHPS1/SIRP/SIRP-ALPHA-1/SIRPalpha/SIRPalpha2 | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse | |
Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.