Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IA2, Polyclonal, Invitrogen™
Chicken Polyclonal Antibody
Supplier: Thermo Scientific PA127627
Description
Implicated in neuroendocrine secretory processes. May be involved in processes specific for neurosecretory granules. such as their biogenesis. trafficking or regulated exocytosis or may have a general role in neuroendocrine functions. Seems to lack intrin
PA1-27627 detects PTPRN from human samples.PA1-27627 may be successfully used in Western blot applications. By Western blot, this antibody has only been tested on fusion protein.The immunogen is a synthetic peptide corresponding to human Receptor-type tyrosine-protein phosphatase.
Specifications
PTPRN | |
Polyclonal | |
PTPRN | |
FLJ16131, IA-2, IA-2/PTP, IA2, ICA512, R-PTP-N | |
Chicken | |
Antigen affinity chromatography | |
RUO | |
5798 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Purified |
Western Blot | |
Unconjugated | |
Q16849 | |
PTPRN | |
Synthetic peptide corresponding to human Receptor-type tyrosine-protein phosphatase. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgY |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction