Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-PTPRT Clone: 1F7,
Mouse Monoclonal Antibody
Supplier: MilliporeSigma™ MABS1158
Description
Specifically detects PTPRT Clone: 1F7 in Human, Mouse, Rat samples, and it is validated for Western Blotting
Receptor-type tyrosine-protein phosphatase T (EC 3.1.3.48; UniProt O14522; also known as R-PTP-T, Receptor protein tyrosine phosphatase, Receptor-type tyrosine-protein phosphatase rho, RPTP-rho, RPTPrho) is encoded by the PTPRT (also known as KIAA0283) gene (Gene ID 11122) in human. RPTPrho/PTPRT is a member of the R2B subfamily of receptor-type protein tyrosine phosphatases (R-PTPs). Known RPTPrho/PTPRT substrates include BCR (Tyr177), paxillin (Tyr188), and STAT3 (Tyr705). In addition, in vitro pull-downs and cell-free dephosphorylation assays also suggest cadherin and catenin family of cell adhesion molecules as physiological substrates of RPTPrho/PTPRT. PTPRT is the single most commonly mutated PTPR gene in all sequenced human cancers, with the highest PTPRT mutation frequency found in cutaneous melanoma. Among 16 tumor types examined, 37.9% of PTPRT mutations are found in the catalytic (PTPase) domain and 33.0% in the extracellular fibronectin type III-like (FN3) domain. In head and neck squamous cell carcinoma (HNSCC), 45.5% PTPRT mutations are located in the PTPase domain, leading to up-regulated STAT3 phosphorylation in HNSCC tumors.
Specifications
PTPRT | |
Monoclonal | |
Unconjugated | |
O14522 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Clone 1F7 reacts with the full-length PTPRT as well as cleaved/truncated PTPRT forms that contain the two phosphatase domains. | |
2°C to 8°C for one year from date of shipment | |
IgG1 κ |
Western Blot | |
1F7 | |
Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
PTPRT, KIAA0283 | |
GST-tagged recombinant protein corresponding to the PTPase domains 1/2 of human PTPRT. | |
100 μg | |
Signaling | |
NP_008981 | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction