Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-PTRF/Cavin-1, Polyclonal

Catalog No. ABE1953MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
ABE1953MI 100 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABE1953MI Supplier MilliporeSigma™ Supplier No. ABE1953
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects PTRF/Cavin-1 Clone: in Human, Rat samples, and it is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blotting

Polymerase I and transcript release factor (UniProt Q6NZI2; also known as Cavin-1, PTRF, RNA polymerase I and transcript release factor, TTF-I interacting peptide 12) is encoded by the PTRF (also known as CGL4, CAVIN, CAVIN1, FKSG13) gene (Gene ID 284119) in human. Cavin-1 is the founding member of the PTRF/Cavin family of cytoplasmic proteins whose expressions are essential for caveola formation. PTRF/Cavin-1, serum deprivation response (SDR)/Cavin-2, SDR-related gene product that binds to C kinase (SRBC)/Cavin-3, and muscle-restricted coiled-coil protein (MURC)/Cavin-4, form an oligomeric assembly termed Cavin complex in the cytosol and associate with caveolin at plasma membrane caveolae. Research shows an essential role for PTRF/Cavin-1 but not the other family members in caveola formation and recruitment of the cavin complex to the plasma membrane. In prostate cancer PC3 cells and during development of zebrafish notochord, lack of PTRF-Cavin expression correlates with lack of caveolae, where caveolin displays increased lateral mobility on the plasma membrane and accelerated lysosomal degradation. Expression of Cavin-1 restores caveolae formation in PC3 cells. Likewise the absence of cavin-1 is also observed in advanced prostate carcinoma. PTRF gene mutations are known to cause congenital generalized lipodystrophy 4 (CGL4).
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTRF/Cavin-1
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. Q6NZI2
Gene Symbols PTRF, FKSG13
Host Species Rabbit
Immunogen Linear peptide corresponding to the N-terminus of human PTRF/Cavin-1.
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Epigenetics, Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_036364
Test Specificity Expected to react with spliced isoforms 1 and 2, but not isoform 3.
Target Species Human, Rat
Content And Storage 2°C to 8°C for one year
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.