Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

polymerase I and transcript release factor, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89018702 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-018-702 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-018-702 Supplier Abnova Corporation Supplier No. H00284119D01P
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human PTRF protein.

Sequence: MEDPTLYIVERPLPGYPDAEAPEPSSAGAQAAEEPSGAGSEELIKSDQVNGVLVLSLLDKIIGAVDQIQLTQAQLEERQAEMEGAVQSIQGELSKLGKAHATTSNTVSKLLEKVRKVSVNVKTVRGSLERQAGQIKKLEVNEAELLRRRNFKVMIYQDEVKLPAKLSISKSLKESEALPEKEGEELGEGERPEEDAAALELSSDEAVEVEEVIEESRAERIKRSGLRRVDDFKKAFSKEKMEKTKVRTRENLEKTRLKTKENLEKTRHTLEKRMNKLGTRLVPAERREKLKTSRDKLRKSFTPDHVVYARSKTAVYKVPPFTFHVKKIREGQVEVLKATEMVEVGADDDEGGAERGEAGDLRRGSSPDVHALLEITEESDAVLVDKSDSD

Specifications

Antigen polymerase I and transcript release factor
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human PTRF protein.
Formulation PBS with no preservative; pH 7.4
Gene PTRF
Gene Accession No. NM_012232.2
Gene Alias FKSG13
Gene Symbols PTRF
Host Species Rabbit
Immunogen PTRF (NP_036364.2, 1 a.a. ∼ 390 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 284119
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.