Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

pituitary tumor-transforming 1 interacting protein, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89016735 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-016-735 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-016-735 Supplier Abnova Corporation Supplier No. H00000754B01P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human PTTG1IP protein.

The encoded protein, which directly binds to pituitary tumor-transforming gene 1 protein (PTTG1), facilitates the nuclear translocation of PTTG1 and potentiates the transcriptional activation of basic fibroblast growth factor by PTTG1. The gene product localizes to both the cytoplasm and nucleus. Its NLS is required for its own nuclear localization, the nuclear localization of PTTG1, and its interaction with PTTG1. [provided by RefSeq

Sequence: MAPGVARGPTPYWRLRLGGAALLLLLIPVAAAQEPPGAACSQNTNKTCEECLKNVSCLWCNTNKACLDYPVTSVLPPASLCKLSSARWGVCWVNFEALIITMSVVGGTLLLGIAICCCCCCRRKRSRKPDRSEEKAMREREERRIRQEERRAEMKTRHDEIRKKYGLFKEENPYARFENN

Specifications

Antigen pituitary tumor-transforming 1 interacting protein
Applications Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human PTTG1IP protein.
Formulation PBS with no preservative; pH 7.4
Gene PTTG1IP
Gene Accession No. BC020983
Gene Alias C21orf1/C21orf3/PBF
Gene Symbols PTTG1IP
Host Species Mouse
Immunogen PTTG1IP (AAH20983, 1 a.a. ∼ 180 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 754
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.