Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ptx3/PITX3 Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB5722
Description
Anti-Ptx3/PITX3 Antibody detects level of Ptx3/PITX3 and has been published and validated for use in western blot, immunohistochemistry.
Specifications
Ptx3/PITX3 | |
Polyclonal | |
Precipitated antibody in a solution of 50% saturated ammonium sulfate and PBS containing no preservatives. | |
Rabbit | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat |
Immunohistochemistry, Western Blot | |
Unconjugated | |
PTX3; TSG14; pentraxin-3; TSG-14; TNFAIP5 | |
Synthetic peptide | |
100 μg | |
NM_002852.2 | |
-70°C for up to 6 months, Avoid Freeze/Thaw Cycles |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction