Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PU.1 Mouse anti-Human, Unlabeled, Clone: G148-74, BD

Catalog No. BDB554268 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB554268 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB554268 Supplier BD Biosciences Supplier No. 554268
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PU.1 is an ets-related transcription factor expressed in B lymphocytes and macrophages. It was initially identified as the oncogene Spi-1 which was found to block erythroblast differentiation. PU.1 functions by binding transcriptional control elements such as μB in the immunoglobulin heavy chain enhancer and π adjacent to μE2. It has been shown to form a complex with NF-EM5, a B cell transcription factor, resulting in the activation of the immunoglobulin κ 3' enhancer. PU.1 also appears to be critically involved in the control of monocyte development by regulating the expression of the macrophage colony-stimulating factor receptor. PU.1 runs as a 40-42kDa protein on SDS-PAGE. G148-74 recognizes human PU.1, an Ets-related transcription factor. PU.1 is expressed in B lymphocytes and macrophages and like other Ets-related proteins, binds to consensus sites in DNA through an 85 amino acid Ets domain in the carboxyl terminal region of the protein. The antibody was raised against a bacterially expressed GST-PU.1 fusion protein.

Host Species: Mouse
Clone: G148-74
Isotype: IgG2a
Species Reactivity: Human
Immunogen: PU.1 Fusion Protein
Formula Weight [Chemical]: 40-42kDa

Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen PU.1
Applications Western Blot
Classification Monoclonal
Clone G148-74
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen PU.1 Fusion Protein
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgG2a
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.