Learn More
PUF60, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human PUF60 peptide using ARM Technology.
Supplier: Abnova Corporation H00022827K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
The protein encoded by this gene is a Ro RNP-binding protein. It interacts with Ro RNPs and their interaction is thought to represent a gain of function for Ro RNPs. This protein also forms a ternary complex with far upstream element (FUSE) and FUSE-binding protein. It can repress a c-myc reporter via the FUSE. It is also known to target transcription factor IIH and inhibit activated transcription. This gene is implicated in the xeroderma pigmentosum disorder. There are two alternatively spliced transcript variants of this gene encoding different isoforms. There seems to be evidence of multiple polyadenylation sites for this gene. [provided by RefSeq
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
PUF60 | |
Monoclonal | |
Unconjugated | |
In 1X PBS, pH 7.4 | |
PUF60 | |
PUF60 | |
A synthetic peptide of human PUF60 | |
RUO | |
22827 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
ELISA, Western Blot | |
1mg | |
poly-U binding splicing factor 60kDa | |
PUF60 | |
FIR, FLJ31379, RoBPI, SIAHBP1 | |
Rabbit | |
1 mg x up to 3 Abs | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.