Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

purine-rich element binding protein A, Mouse, Clone: 3A9, Abnova™

Catalog No. 89015839 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-015-839 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-015-839 Supplier Abnova Corporation Supplier No. H00005813M05
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant PURA.

This gene product is a sequence-specific, single-stranded DNA-binding protein. It binds preferentially to the single strand of the purine-rich element termed PUR, which is present at origins of replication and in gene flanking regions in a variety of eukaryotes from yeasts through humans. Thus, it is implicated in the control of both DNA replication and transcription. Deletion of this gene has been associated with myelodysplastic syndrome and acute myelogenous leukemia. [provided by RefSeq

Sequence: TQGQTIALPAQGLIEFRDALAKLIDDYGVEEEPAELPEGTSLTVDNKRFFFDVGSNKYGVFMRVSEVKPTYRNSITVPYKVWAKFGHTFCKYSEETKKIQEKQREKRAAC

Specifications

Antigen purine-rich element binding protein A
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3A9
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant PURA.
Formulation PBS with no preservative; pH 7.4
Gene PURA
Gene Accession No. NM_005859
Gene Alias PUR-ALPHA/PUR1/PURALPHA
Gene Symbols PURA
Host Species Mouse
Immunogen PURA (NP_005850, 183 a.a. ∼ 292 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Cycle
Primary or Secondary Primary
Gene ID (Entrez) 5813
Target Species Human, Rat
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.