Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Puromycin Alexa Fluor™ 488 Conjugate Mouse anti-Human, AlexaFluor™488, Clone: 12D10, MilliporeSigma™

Catalog No. MABE343AF48 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE343AF48 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE343AF48 Supplier MilliporeSigma Supplier No. MABE343AF488
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-Puromycin, clone 12D10 is validated for use in Immunocytochemistry for the detection of Puromycin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Puromycin
Applications Immunocytochemistry
Classification Monoclonal
Clone 12D10
Concentration 0.5mg/mL
Conjugate Alexa Fluor 488
Formulation Purified rat monoclonal conjugate in buffer containing PBS and 15mg/mL BSA with 0.1% Sodium Azide.
Host Species Mouse
Immunogen Puromycin-incorporated neosynthesized proteins at multiple molecular weights
Purification Method Protein G purified
Quantity 100 μL
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.