Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Puromycin Mouse anti-Human, All, Clone: 4G11, MilliporeSigma™

Catalog No. MABE342MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE342MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE342MI Supplier MilliporeSigma Supplier No. MABE342
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Puromycin antibody, clone 4G11 detects puromycin incorporated into protein. Monoclonal antibodies to puromycin may also be used with standard immunochemical methods.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Puromycin
Applications Immunocytochemistry, Immunofluorescence, Western Blot, Western Blot
Classification Monoclonal
Clone 4G11
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Host Species Mouse
Immunogen Puromycin from Streptomyces alboniger
Purification Method Protein G purified
Quantity 100 μL
Primary or Secondary Primary
Target Species All species, Human
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.