Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Puromycin Sensitive Aminopeptidase Goat anti-Mouse, Rat, Polyclonal, MilliporeSigma™

Catalog No. AB3258MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB3258MI 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB3258MI Supplier MilliporeSigma Supplier No. AB3258
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This Anti-Puromycin Sensitive Aminopeptidase Antibody is validated for use in western blot, IC for the detection of Puromycin Sensitive Aminopeptidase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Puromycin Sensitive Aminopeptidase
Applications Immunocytochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified immunoglobulin. Liquid in phosphate buffered saline.
Gene Symbols NPEPPS; MP100; PSA; EC 3.4.11.-
Host Species Goat
Immunogen Rat puromycin sensitive aminopeptidase.
Quantity 50 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_006310.3
Target Species Mouse, Rat
Content And Storage −20°C in undiluted aliquotes for 6 months. Avoid repeated freeze/thaw cycles.
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.