Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PYGM Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA511511
Description
This antibody is predicted to react with non-human primate based on sequence homology.
PYGM catalyzes and regulates the breakdown of glycogen to glucose-1-phosphate. Defects in PYGM are the cause of glycogen storage disease type 5 (GSD5), also known as McArdle disease. GSD5 is a metabolic disorder resulting in myopathy characterized by exercise intolerance, cramps, muscle weakness and recurrent myoglobinuria.
Specifications
PYGM | |
Polyclonal | |
PYGM | |
Glycogen phosphorylase; muscle form; Myophosphorylase; PYGM; | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
5837 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
P11217 | |
PYGM | |
KLH conjugated synthetic peptide between 698-727 amino acids from the C-terminal region of human PYGM | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction