Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PYY Chicken anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™

Catalog No. AB15666MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB15666MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB15666MI Supplier MilliporeSigma Supplier No. AB15666
Add to Cart
Add to Cart

Chicken Polyclonal Antibody

This Anti-PYY Antibody, amino acids 24-36 is validated for use in ELISA for the detection of PYY.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PYY
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
Formulation Liquid in PBS with 0.02% sodium azide.
Gene Symbols PYY; PYY1; PYY-I
Host Species Chicken
Immunogen Synthetic peptide from the C-terminus of human PYY (amino acids 24-36).
Purification Method Affinity Purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_004160.3
Target Species Human, Mouse, Rat
Content And Storage −20°C in undiluted aliquotes for 6 months. Avoid repeated freeze/thaw cycles.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.