Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
QKI-5 Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB9904
Description
Detect QKI-5 using this Anti-QKI-5 Antibody validated for use in immunoprecipitation and western blotting.
Specifications
QKI-5 | |
Polyclonal | |
Liquid. | |
Rabbit | |
Serum | |
Primary | |
Human, Mouse, Rat | |
Serum |
Immunoprecipitation, Western Blot | |
Unconjugated | |
QKI; DKFZp586I0923; Hqk; QK3; QK; HKQ; HqkI | |
Synthetic peptide from mouse QKI-5. | |
100 μL | |
NM_006775.1; NM_206853.1; NM_206854.1; NM_206855.1 | |
−20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction