Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

QKI-6 Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™

Catalog No. AB9906MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB9906MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB9906MI Supplier MilliporeSigma Supplier No. AB9906
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-QKI-6 Antibody detects level of QKI-6 and has been published and validated for use in immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen QKI-6
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Liquid.
Gene Symbols QKI; DKFZp586I0923; Hqk; QK3; QK; HKQ; HqkI
Host Species Rabbit
Immunogen Synthetic peptide from mouse QKI-6.
Purification Method Serum
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_006775.1; NM_206853.1; NM_206854.1; NM_206855.1
Target Species Human, Mouse, Rat
Content And Storage −20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles.
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.