Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

quinolinate phosphoribosyltransferase (D05P), Rabbit anti-Human, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89018100 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-018-100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-018-100 Supplier Abnova Corporation Supplier No. H00023475D05P
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human QPRT protein.

This gene encodes a key enzyme in catabolism of quinolinate, an intermediate in the tryptophan-nicotinamide adenine dinucleotide pathway. Quinolinate acts as a most potent endogenous exitotoxin to neurons. Elevation of quinolinate levels in the brain has been linked to the pathogenesis of neurodegenerative disorders such as epilepsy, Alzheimer's disease, and Huntington's disease. [provided by RefSeq]

Sequence: MDAEGLALLLPPVTLAALVDSWLREDCPGLNYAALVSGAGPSQAALWAKSPGVLAGQPFFDAIFTQLNCQVSWFLPEGSKLVPVARVAEVRGPAHCLLLGERVALNTLARCSGIASAAAAAVEAARGAGWTGHVAGTRKTTPGFRLVEKYGLLVGGAASHRYDLGGLVMVKDNHVVAAGGVEKAVRAARQAADFALKVEVECSSLQEAVQAAEAGADLVLLDNFKPEELHPTATVLKAQFPSVAVEASGGITLDNLPQFCGPHIDVISMGMLTQAAPALDFSLKLFAKEVAPVPKIH

Specifications

Antigen quinolinate phosphoribosyltransferase
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human QPRT protein.
Formulation PBS with no preservative; pH 7.4
Gene QPRT
Gene Accession No. NM_014298.3
Gene Alias QPRTase
Gene Symbols QPRT
Host Species Rabbit
Immunogen QPRT (NP_055113.2, 1 a.a. ∼ 297 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23475
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.