Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

QTRT1, Rabbit, Monoclonal Antibody, Abnova™

Catalog No. 89208759 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg x up to 3 Abs
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-208-759 1 mg x up to 3 Abs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-208-759 Supplier Abnova Corporation Supplier No. H00081890K

Includes

Up to three rabbit IgG clones of 1mg each will be delivered to customer.

Rabbit monoclonal antibody raised against a human QTRT1 peptide using ARM Technology.

tRNA-guanine transglycosylase (TGT; EC 2.4.2.29) synthesizes queuosine (Q), which is found in tRNAs that recognize NAU and NAC codons, encoding tyr, asn, asp, and his. Prokaryotic TGT is a single protein of 43kDa. In contrast, mammalian TGT appears to be a heterodimer consisting of a 60kDa subunit (USP14; MIM 607274) and a 43kDa catalytic subunit (QTRT1) (Deshpande and Katze, 2001 [PubMed 11255023]).[supplied by OMIM

Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)
Expression: Overexpression vector and transfection into 293H cell line

Order Info

  1. Customer may provide cell or tissue lysate for antibody screening.
  2. Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.

Specifications

Antigen QTRT1
Applications ELISA, Western Blot
Classification Monoclonal
Concentration 1mg
Conjugate Unconjugated
Description queuine tRNA-ribosyltransferase 1
Formulation In 1X PBS, pH 7.4
Gene QTRT1
Gene Accession No. QTRT1
Gene Alias FP3235, TGT
Gene Symbols QTRT1
Host Species Rabbit
Immunogen A synthetic peptide of human QTRT1
Quantity 1 mg x up to 3 Abs
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81890
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.