Learn More
R-Phycoerythrin Mouse, Biotin, Clone: E31-1459, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 552603
Description
The E31-1459 antibody reacts with R-Phycoerythrin (PE), a commonly used fluorochrome for flow cytometry, alone or as a tandem conjugate with other fluorochromes (eg, PE-Cy™7). The binding of the E31-1459 antibody to PE does not quench the fluorescence of the PE molecule. The use of biotinylated E31-1459 in a three-step staining procedure with Streptavidin-PE and a PE-conjugated primary antibody can specifically amplify the PE fluorescence.
Host Species: Mouse
Clone: E31-1459
Isotype: IgG1 κ
Immunogen: R-phycoerythrin
Flow Cytometry

Specifications
R-Phycoerythrin | |
Monoclonal | |
0.5mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
R-phycoerythrin | |
0.1 mg | |
Immunology | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
E31-1459 | |
Biotin | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.