Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rab11 Mouse, Unlabeled, Clone: 47, BD

Catalog No. BDB610657 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610657 150 μg
BDB610656 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610657 Supplier BD Biosciences Supplier No. 610657
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The Rab proteins are small GTP-binding molecules that are localized to specific intracellular vesicles and organelles. It has been proposed that Rab proteins cycle between GTP- and GDP-bound forms and that this is related to their function as regulators of vesicular traffic. The Rab11 gene encodes a 24 kDa protein of 214 amino acids that has been detected in liver, brain, testis, spleen, and heart. Rab11 protein was isolated from the golgi-microsomal fraction of rat liver and has been detected in the Trans-golgi Network, secretory vesicles, and the pericentriolar recycling endosomes. The distribution of Rab11 indicates that this small protein is involved in regulating traffic at the Golgi complex.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Rab11
Applications Western Blot
Classification Monoclonal
Clone 47
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Purification Method Affinity Purified
Quantity 150 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Chicken, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG2a
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.