Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAB11A, member RAS oncogene family, Mouse, Clone: 1F7-2A8, Abnova™

Catalog No. 89014225 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-014-225 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-014-225 Supplier Abnova Corporation Supplier No. H00008766M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant RAB11A.

The protein encoded by this gene belongs to the small GTPase superfamily, Rab family. It is associated with both constitutive and regulated secretory pathways, and may be involved in protein transport. [provided by RefSeq

Sequence: MGTRDDEYDYLFKVVLIGDSGVGKSNLLSRFTRNEFNLESKSTIGVEFATRSIQVDGKTIKAQIWDTAGQERYRAITSAYYRGAVGALLVYDIAKHLTYENVERWLKELRDHADSNIVIMLVGNKSDLRHLRAVPTDEARAFAEKNGLSFIETSALDSTNVEAAFQTILTEIYRIVSQKQMSDRRENDMSPSNNVVPIHVPPTTENKPKVQCCQNI

Specifications

Antigen RAB11A, member RAS oncogene family
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1F7-2A8
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full length recombinant RAB11A.
Formulation PBS with no preservative; pH 7.4
Gene RAB11A
Gene Accession No. BC013348
Gene Alias MGC1490/YL8
Gene Symbols RAB11A
Host Species Mouse
Immunogen RAB11A (AAH13348.1, 1 a.a. ∼ 216 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8766
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.