Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAB11B, member RAS oncogene family, Mouse, Clone: 2F1, Abnova™

Catalog No. 89001507 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-001-507 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-001-507 Supplier Abnova Corporation Supplier No. H00009230M03A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant RAB11B.

Sequence: MGTRDDEYDYLFKVVLIGDSGVGKSNLLSRFTRNEFNLESKSTIGVEFATRSIQVDGKTIKAQIWDTAGQERYRAITSAYYRGAVGALLVYDIAKHLTYENVERWLKELR

Specifications

Antigen RAB11B, member RAS oncogene family
Applications ELISA
Classification Monoclonal
Clone 2F1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant RAB11B.
Formulation ascites with no preservative
Gene RAB11B
Gene Accession No. NM_004218
Gene Alias H-YPT3/MGC133246
Gene Symbols RAB11B
Host Species Mouse
Immunogen RAB11B (NP_004209, 1 a.a. ∼ 110 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Research Discipline Signal Transduction
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 9230
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.