Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RAB11B, member RAS oncogene family, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human RAB11B protein.
Supplier: Abnova Corporation H00009230B02P
Description
Sequence: MGTRDDEYDYLFKVVLIGDSGVGKSNLLSRFTRNEFNLESKSTIGVEFATRSIQVDGKTIKAQIWDTAGQERYRRITSAYYRGAVGALLVYDIAKHLTYENVERWLKELRDHADSNIVIMLVGNKSDLRHLRAVPTDEARAFAEKNNLSFIETSALDSTNVEEAFKNILTEIYRIVSQKQIADRAAHDESPGNNVVDISVPPTTDGQKPNKLQCCQNLSpecifications
RAB11B, member RAS oncogene family | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human RAB11B protein. | |
RAB11B | |
H-YPT3/MGC133246 | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
Antibody |
Western Blot | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_004218 | |
RAB11B | |
RAB11B (NP_004209, 1 a.a. ∼ 218 a.a) full-length human protein. | |
50 μg | |
Signal Transduction | |
9230 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction