Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rab13 Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™

Catalog No. 07794MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-794-MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-794-MI Supplier MilliporeSigma Supplier No. 07794
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Detect Rab13 using this Anti-Rab13 Antibody validated for use in western blot, immunoprecipitation and immunofluoresence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rab13
Applications Immunofluorescence, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Affinity Purified rabbit IgG in storage buffer containing tri-citrate-phosphate, pH 7-8 and 0.1% sodium azide.
Gene Symbols Rab13; RAB13, member RAS oncogene family ; GIG4; growth-inhibiting gene 4 protein
Host Species Rabbit
Immunogen Sythnetic peptide of amno acids 129-148 of rat Rab13
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NP_002861
Target Species Human, Mouse, Rat
Content And Storage 2-8°C for 12 months
Handling Recommendations: Upon receipt and prior to removing the cap centrifuge the vial and gently mix the solution. Avoid repeated freeze/thaw cycles which may damage IgG and affect product performance.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.