Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-Rab18, Polyclonal

Catalog No. 55212650UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
55-212-650UL 50 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 55-212-650UL Supplier MilliporeSigma™ Supplier No. 55212650UL

Rabbit Polyclonal Antibody

Specifically detects Rab18 Clone: in Rat samples, and it is validated for ELISA, Immuno Blotting

A small GTP-binding protein of the Rab family that is involved in vesicular transport. It is localized on the apical side of epithelial cells and may play a role in endocytosis and membrane recycling.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rab18
Applications ELISA, Immunoblot, Immunoblot
Classification Polyclonal
Conjugate Unconjugated
Formulation Undiluted serum.
Host Species Rabbit
Immunogen recombinant rat brain Rab18
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the Rab18 protein.
Target Species Rat
Content And Storage −20°C
Form Undiluted
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.