Learn More
RAB23, member RAS oncogene family, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human RAB23 protein.
Supplier: Abnova Corporation H00051715D01P
Description
The protein encoded by this gene belongs to the small GTPase superfamily, Rab family. It may be involved in small GTPase mediated signal transduction and intracellular protein transportation. Alternative splicing occurs at this locus and two transcript variants encoding the same protein have been identified. [provided by RefSeq
Sequence: MLEEDMEVAIKMVVVGNGAVGKSSMIQRYCKGIFTKDYKKTIGVDFLERQIQVNDEDVRLMLWDTAGQEEFDAITKAYYRGAQACVLVFSTTDRESFEAVSSWREKVVAEVGDIPTVLVQNKIDLLDDSCIKNEEAEALAKRLKLRFYRTSVKEDLNVNEVFKYLAEKYLQKLKQQIAEDPELTHSSSNKIGVFNTSGGSHSGQNSGTLNGGDVINLRPNKQRTKKNRNPFSSCSIPSpecifications
| RAB23, member RAS oncogene family | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against a full-length human RAB23 protein. | |
| RAB23 | |
| DKFZp781H0695/HSPC137/MGC8900 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Antibody |
| Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_016277 | |
| RAB23 | |
| RAB23 (NP_057361.3, 1 a.a. ∼ 237 a.a) full-length human protein. | |
| 100 μg | |
| Signal Transduction | |
| 51715 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.