Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAB27 Mouse anti-Human, Unlabeled, Clone: 20.1/RAB27, BD

Catalog No. BDB558532 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB558532 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB558532 Supplier BD Biosciences Supplier No. 558532
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Members of the Rab family of GTPases are non-transforming, membrane-bound, geranylgeranylated members of the Ras superfamily that are involved in the fusion and trafficking of cellular vesicles. Rab27A is expressed in many human tissues and it regulates the secreation of several types of cellular vesicles, such as melanosomes, insulin granules of pancreatic beta cells, and lytic granules of cytotoxic T lymphocytes. Rab27A is able to regulate such diverse processes by interacting with organelle-specific effector proteins, including melanophilin, granuphilin, and Munc13-4, respectively. Defects in Rab27A function are associated with the human diseases choroideremia and Griscelli syndrome 2 and the ashen mutant mouse. Rab27B is not so widely expressed, yet it is reported to have similar functional capacities as Rab27A. The 20.1 monoclonal antibody recognizes human Rab27A; cross-reactivity with Rab27B is unknown.

Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen RAB27
Applications Western Blot
Classification Monoclonal
Clone 20.1/RAB27
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human Rab27A Recombinant Protein
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.