Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RAB3IL1 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA524512
Description
Guanine nucleotide exchange factor (GEF) for Rab3A, a GTPase that regulates synaptic vesicle exocytosis.
Specifications
RAB3IL1 | |
Polyclonal | |
RAB3IL1 | |
GRAB, guanine nucleotide exchange factor for Rab-3A, guanine nucleotide exchange factor for Rab3A, rab-3A-interacting-like protein 1, rab3A-interacting-like protein 1, rabin3-like 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5866 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Western Blot | |
Unconjugated | |
Q8TBN0 | |
RAB3IL1 | |
KLH conjugated synthetic peptide between 297-325 amino acids from the C-terminal region of human RAB3IL1 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction