Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rab4 Rabbit anti-Canine, Chicken, Guinea Pig, Human, Hamster, Mouse, Monkey, Pig, Rat, Rabbit, Sheep, Polyclonal, MilliporeSigma™

Catalog No. 07655MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-655-MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-655-MI Supplier MilliporeSigma Supplier No. 07655
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Rab4 Antibody detects level of Rab4 and has been published and validated for use in immunoprecipitation, western blotting and immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rab4
Applications Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Gene Symbols RAB4A; RAB4; OTTHUMP00000042824
Host Species Rabbit
Immunogen GST fusion protein corresponding to human Rab4.
Purification Method Serum
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_004578.2
Target Species Canine, Chicken, Guinea Pig, Hamster, Human, Monkey, Mouse, Pig, Rabbit, Rat, Sheep
Content And Storage −20°C for 24 months
Form Serum
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.