Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rab40C Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™

Catalog No. 071392MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-139-2MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-139-2MI Supplier MilliporeSigma Supplier No. 071392
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Use Anti-Rab40C Antibody (Rabbit Polyclonal Antibody) validated in western blot, immunohistochemistry to detect Rab40C also known as RAS-like GTPase, SOCS box-containing protein RAR3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rab40C
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Rabbit Serum
Gene Symbols RARL; RASL8C; RAB40C
Host Species Rabbit
Immunogen Full-length His-tagged Rab40C recombinant protein (mouse).
Purification Method Unpurified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NP_066991
Target Species Human, Mouse, Rat
Content And Storage −20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.