Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rab5 Mouse, Unlabeled, Clone: 1, BD

Catalog No. BDB610725 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610725 150 μg
BDB610724 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610725 Supplier BD Biosciences Supplier No. 610725
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Rab5 is a low molecular weight GTP-binding protein that plays a role in endocytic vesicle traffic. Like other Rab proteins, Rab5 has C-terminal cysteine residues that are post translationally modified by geranylgeranylation which is critical for its membrane targeting. Rab5 is associated with early endosome and plasma membranes and evidence suggests that Rab5 regulates early endosome fusion. The GTP/GDP cycle controls shuttling of Rab proteins between the cytosol and membranes. In vitro, Rab5 proteins are removed from membranes by a GDP dissociation inhibitor protein (rabGDI) which leads to the formation of a cytosolic Rab5-rabGDI complex. Rab5 may insert into membranes by a multistep process in a which a transient GDP-Rab5 intermediate is formed and converted into GTP-Rab5 that subsequently enters the acceptor membrane and releases rabGDI into the cytosol.

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Rab5
Applications Western Blot
Classification Monoclonal
Clone 1
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human Rab5 aa. 1-215
Purification Method Affinity Purified
Quantity 150 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Human
Content And Storage Store undiluted at -20°C.
Isotype IgG2a
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.