Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-Rab5, Polyclonal

Catalog No. 55210850UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
55-210-850UL 50 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 55-210-850UL Supplier MilliporeSigma™ Supplier No. 55210850UL

Rabbit Polyclonal Antibody

Specifically detects Rab5 Clone: in Human samples, and it is validated for ELISA, Immunohistochemistry, Immuno Blotting

Rab5 is a small GTPase that regulates the fusion of endocytic vesicles to early endosomes. It is also involved in axonal and dendritic endocytosis and corresponds to yeast Ypt51.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rab5
Applications ELISA, Immunoblot, Immunoblot, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation Undiluted serum.
Host Species Rabbit
Immunogen recombinant, human retinal Rab5A
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the Rab5 protein.
Target Species Human
Content And Storage −20°C
Form Undiluted
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.