Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAB5A, member RAS oncogene family, Mouse, Clone: 5D1, Abnova™

Catalog No. 89001481 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-001-481 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-001-481 Supplier Abnova Corporation Supplier No. H00005868M09A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant RAB5A.

Sequence: KELQRQASPNIVIALSGNKADLANKRAVDFQEAQSYADDNSLLFMETSAKTSMNVNEIFMAIAKKLPKNEPQNPGANSARGRGVDLTEPTQPTRNQCCSN

Specifications

Antigen RAB5A, member RAS oncogene family
Applications ELISA, Western Blot
Classification Monoclonal
Clone 5D1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant RAB5A.
Formulation ascites with no preservative
Gene RAB5A
Gene Accession No. BC001267
Gene Alias RAB5
Gene Symbols RAB5A
Host Species Mouse
Immunogen RAB5A (AAH01267, 116 a.a. ∼ 215 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Research Discipline Signal Transduction
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 5868
Target Species Human, Mouse
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.