Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rab8 Mouse, Unlabeled, Clone: 4, BD

Catalog No. BDB610844 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610844 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB610844 Supplier BD Biosciences Supplier No. 610844
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The members of the Ras superfamily of GTPases, which now includes more than 50 different proteins, shuttle between the GDP-bound inactive position and the GTP-bound active state. The GTPase activity of the Ras-like proteins is regulated by effectors that promote either the GDP- or the GTP-bound states. GTPases of the Ras superfamily have a role in controlling numerous events such as cell proliferation, nucleocytoplasmic transport, and vesicular traffic. The Rab superfamily of proteins includes more than 30 different members that are involved in the regulation of different steps of vesicular traffic. Subcellular localization and specific cellular function of each Rab appears to be encoded in the unique C terminal part of the proteins, known as the hypervariable region. The 207 amino acid Rab8 protein is closely related to the yeast Sec4p and regulates vesicle transport from the Trans-golgi Network to the basolateral plasma membrane in epithelial cells. In addition, Rab8 interacts with the stress-activated kinase rab8p, a close homologue of the GC Kinase. This interaction was found to be dependent on GTP.

Immunofluorescence, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Rab8
Applications Western Blot
Classification Monoclonal
Clone 4
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human Rab8 aa. 84-205
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Drosophila, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG2b
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.