Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RAB8A Rabbit anti-Human, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma ABT165
Description
This Anti-RAB8A Antibody is validated for use in Western Blotting for the detection of RAB8A.
Specifications
RAB8A | |
Polyclonal | |
Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
Rabbit | |
Affinity Purified | |
Primary | |
Human |
Western Blot | |
Unconjugated | |
RAB8A; MEL; RAB8 | |
KLH-conjugated linear peptide corresponding to the C-terminus of human RAB8A. | |
100 μg | |
NP_005361 | |
2-8°C for 12 months |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction