Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit IgG Isotype Control, Novus Biologicals™
SDP

Catalog No. NBP236463 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP236463 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP236463 Supplier Novus Biologicals Supplier No. NBP236463
Add to Cart
Add to Cart

Rabbit Isotype Control Antibody

IgG Isotype Control antibody specifically detects IgG in Rabbit samples. It is validated for Western Blot, ChIP assay, ELISA, Immunohistochemistry, Flow Cytometry.

Specifications

Antigen IgG
Applications Western Blot, ELISA, Immunohistochemistry, Flow Cytometry, ChIP Assay
Concentration 10.0 mg/ml
Conjugate Unconjugated
Dilution Western Blot, ELISA, Immunohistochemistry, SDS-Page, Flow (Intracellular), Chromatin Immunoprecipitation (ChIP)
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2. with 0.01% Sodium Azide
Gene Alias Immunoglobulin G, ImmunoglobulinG
Gene Symbols IGHG1
Host Species Rabbit
Purification Method Multi-step
Quantity 0.5 mg
Regulatory Status RUO
Primary or Secondary Isotype Control (Primary)
Gene ID (Entrez) 3500
Test Specificity Rabbit IgG whole molecular was prepared from normal serum by a multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the buffer. Rabbit IgG whole molecular was assayed by immunoelectrophoresis resulted in a single precipitin arc against anti-Rabbit IgG and anti-Rabbit Serum.
Target Species Rabbit
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Concentrated
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.