Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit IgG Isotype Control, Novus Biologicals™
SDP

Catalog No. NBP224891 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224891 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP224891 Supplier Novus Biologicals Supplier No. NBP224891
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 36 publications

IgG Polyclonal specifically detects IgG in Rabbit samples. It is validated for Chromatin Immunoprecipitation, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, In vitro assay.

Specifications

Antigen IgG
Applications ChIP Assay, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Chromatin Immunoprecipitation 1:10-1:500. Use reported in scientific literature (PMID 23034403), Flow Cytometry 1:10 - 1:1000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 32784470), Immunohistochemistry-Paraffin 1:10 - 1:500. Use reported in scientific literature (PMID 255994242), In vitro assay, Chromatin Immunoprecipitation (ChIP) 1:10-1:500
Formulation PBS with 0.05% Sodium Azide
Gene Alias Immunoglobulin G, ImmunoglobulinG
Gene Symbols IGHG1
Host Species Rabbit
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Isotype Control (Primary)
Gene ID (Entrez) 3500
Target Species Rabbit
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.