Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabex-5 Mouse anti-Mouse, Unlabeled, Clone: 27, BD

Catalog No. BDB612559 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB612559 150 μg
BDB612558 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB612559 Supplier BD Biosciences Supplier No. 612559

Mouse Monoclonal Antibody

The Rab proteins are small GTP-binding molecules. They are localized to specific intracellular vesicles and organelles and are important for vesicular trafficking, cycling between an active GTP-bound and inactive GDP-bound form. Rab5 is associated with vesicle trafficking between the early endosomes and plasma membrane. In vitro, Rab5 proteins are removed from membranes by a GDP dissociation inhibitor protein (rabGDI). This leads to the formation of a cytosolic Rab5-rabGDI complex. Rab5 may insert into membranes by a multistep process in which a transient GDP-Rab5 intermediate is formed and converted into GTP- Rab5. Rabaptin-5 interacts with GTP-Rab5, and is recruited to the endosomal fraction in a Rab5/GTP-dependent manner. Removal of Rabaptin-5 from the cytosol substantially impairs GTP and Rab5-dependent endosomal fusion. Rabex-5 forms a complex with Rabaptin-5 and displays GDP/GTP exchange activity on Rab5 that promotes interaction between Rabaptin-5 and Rab5. Rabex-5 is also known as Rin2 based on its 15% identitiy with the RasGTPase-binding protein Rin1. Thus, Rabex-5/Rabaptin-5 complex is critical for Rab5 GDP/GTP exchange and membrane-associated activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rabex-5
Applications Western Blot
Classification Monoclonal
Clone 27
Concentration 250μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Mouse Rabex-5 aa. 426-481
Purification Method Affinity Purified
Quantity 150 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Mouse
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.