Learn More
RAB, member of RAS oncogene family-like 2B, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human RABL2B protein.
Supplier: Abnova Corporation H00011158B01
Description
The RABL2B protein is a member of the RAB gene family which belongs to the RAS GTPase superfamily. RABL2B is located within a subtelomeric region of 22q13.3. Multiple alternatively spliced transcript variants encoding several different isoforms have been found for this gene. [provided by RefSeq
Sequence: MAEDKTKPSELDQGKYDADDNVKIICLGDSAVGKSKLMERFLMDGFQPQQLSTYALTLYKHTATVDGRTILVDFWDTAGQERFQSMHASYYHKAHACIMVFDVQRKVTYRNLSTWYTELREFRPEIPCIVVANKIDADINVTQKSFNFAKKFSLPLYFVSAADGTNVVKLFNDAIRLAVSYKQNSQDFMDEIFQELENFSLEQEEEDVPDQEQSSSIETPSEEAASPHSSpecifications
RAB, member of RAS oncogene family-like 2B | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human RABL2B protein. | |
RABL2B | |
FLJ93981/FLJ98216 | |
Mouse | |
50 μL | |
Signal Transduction | |
Primary | |
Human | |
Serum |
Western Blot | |
Unconjugated | |
No additive | |
NM_001003789 | |
RABL2B | |
RABL2B (NP_001003789, 1 a.a. ∼ 229 a.a) full-length human protein. | |
RUO | |
Yes | |
11158 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.