Learn More
Rac1 Mouse, Unlabeled, Clone: 102, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 610650
Description
Rac1 is a member of the expanding family of Ras-related GTPase proteins. These small proteins have the ability to switch between two different conformational states: the active (GTP-bound) and the inactive (GDP-bound). The GTPase activity is increased by specific activator proteins named GAPs. Rac1, like many of its relatives, contains the consensus sequence Cys-X-X-X-COOH, which localizes it to the plasma membrane. Growth factor stimulation of cells results in a dramatic reorganization of the actin filament network and membrane ruffling, processes for which Rac1 is essential. Constitutive expression of Rac1 leads to cell transformation. This process is mediated by the Rac1-dependent activation of the p65PAK serine/threonine kinase. In addition, IQGAP1 has been identified as a GTPase activating protein specific for Rac1. Due to sequence homology among various small GTPase family members, potential cross-reactivity could be observed with this antibody. BD Biosciences Pharmingen, however, has not performed such cross-reactivity testing.
This antibody is routinely tested by western blot analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Specifications
| Rac1 | |
| Monoclonal | |
| 250μg/mL | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| 50 μg | |
| Cell Biology | |
| Canine, Chicken, Human, Mouse, Rat | |
| IgG2b κ |
| Western Blot | |
| 102 | |
| Unconjugated | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at -20°C. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.