Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rac1 Mouse anti-Human, Mouse, Rat, Clone: 23A8, MilliporeSigma™

Catalog No. 16319MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-319-MI 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-319-MI Supplier MilliporeSigma Supplier No. 16319
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Rac1 Antibody, clone 23A8, Magnetic bead Conjugate detects level of Rac1 and has been published and validated for use in western blotting and immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rac1
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 23A8
Conjugate Unconjugated
Formulation 20% slurry; 1xPBS, 0.05% sodium azide. Sufficient for 10 immunoprecipitation assays.
Gene Accession No. NP_008839
Gene Symbols RAC1, TC25, MIG5
Host Species Mouse
Immunogen Recombinant protein containing the full length human Rac1.
Quantity 400 μL
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage 2-8°C, Do not Freeze. Wash the magnetic beads with an appropriate buffer prior to use to remove sodium azide.
Form Ascites
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.