Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rac1 Mouse anti-Rat, Mouse, Human, FITC, Clone: 23A8, MilliporeSigma™

Catalog No. FCMAB330FMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FCMAB330FMI 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FCMAB330FMI Supplier MilliporeSigma Supplier No. FCMAB330F

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

Milli-Mark™ Anti-Rac1-FITC Antibody, clone 23A8 detects level of Rac1 and has been published and validated for use in flow cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rac1
Applications Flow Cytometry
Classification Monoclonal
Clone 23A8
Conjugate FITC
Formulation Purified mouse monoclonal IgG2b conjugated to FITC in PBS with 0.1% sodium azide and 15mg/mL BSA.
Gene Accession No. NP_008839
Gene Symbols RAC1, MGC111543, ORFNames=MIG5, PAK1, MIG5, TC-25, p21-Rac1
Host Species Mouse
Immunogen Recombinant protein containing the full length human Rac1.
Purification Method Protein G Purified
Quantity 100 Tests
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage 2-8°C in undiluted aliquots for up to 6 months. Protect from Light.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.