Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rac1 Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™

Catalog No. 071464MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-146-4MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-146-4MI Supplier MilliporeSigma Supplier No. 071464
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Rac1 Antibody is an antibody against Rac1 for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rac1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal IgG in 0.02M PBS with 0.15M NaCl, pH 7.6 and 0.1% sodium azide as a preservative
Gene Symbols RAC1; ORFNames=MIG5; TC-25; p21-Rac1; MGC111543; PAK1; MIG5
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 130-141 of human Rac1 (KEKKLTPITYPQ-C).
Purification Method Protein A purfied
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_006908.4
Target Species Human, Mouse, Rat
Content And Storage 2-8°C in undiluted aliquotes for up to 6 months
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.